Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006261.4 | NP_006252.3 | 3 | 1319 | ||
GenBank mRNA | AF076215.1 | - | 3 | 1319 |