Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015089.3 | NP_055904.1 | 23 | 4567 | ||
XM_011514422.1 | XP_011512724.1 | 23 | 4576 | |||
XM_011514423.2 | XP_011512725.1 | 24 | 4678 | |||
XM_011514424.1 | XP_011512726.1 | 23 | 4495 | |||
XM_011514425.1 | XP_011512727.1 | 21 | 4246 | |||
XM_011514426.1 | XP_011512728.1 | 23 | 4576 | |||
XM_011514427.1 | XP_011512729.1 | 19 | 3195 | |||
XM_011514428.1 | XP_011512730.1 | 23 | 4576 | |||
XM_011514429.1 | XP_011512731.1 | 16 | 2400 | |||
XM_017010589.1 | XP_016866078.1 | 23 | 5287 | |||
XM_017010590.1 | XP_016866079.1 | 17 | 2749 | |||
GenBank mRNA | AB014608.3 | - | 15 | 2201 | ||
AJ318215.1 | - | 22 | 4492 | |||
AY145132.1 | - | 22 | 4492 | |||
CR749511.1 | - | 22 | 4946 | |||
CR749841.1 | - | 23 | 4566 |