Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105677.2 | NP_001099147.2 | 1 | 155 | ||
NM_001301233.1 | NP_001288162.1 | 1 | 155 | |||
GenBank mRNA | FJ664272.1 | - | 1 | 155 | ||
FJ664273.1 | - | 1 | 155 |
Gene Symbol: UGT2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252274.2 | NP_001239203.2 | 3 | 872 | ||
GenBank mRNA | AK304249.1 | - | 3 | 872 |