Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020169.3 | NP_064554.3 | 3 | 570 | ||
XM_017006876.1 | XP_016862365.1 | 3 | 599 | |||
GenBank mRNA | AF087851.1 | - | 3 | 433 | ||
AF282594.1 | - | 3 | 439 | |||
AF282626.1 | - | 3 | 506 | |||
BC005346.1 | - | 3 | 570 | 3932274 | ||
BC008438.1 | - | 3 | 552 | 4093065 |