Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257370.1 | NP_001244299.1 | 3, 4 | 475 | ||
XM_017023025.1 | XP_016878514.1 | 3, 4 | 397 | |||
GenBank mRNA | AK074080.2 | - | 3, 4 | 377 | ||
EF452422.1 | - | 3, 4 | 475 |