Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101.3 | NP_001092.1 | 6 | 1781 | ||
GenBank mRNA | AK025375.1 | - | 6 | 1780 | ||
AK058019.1 | - | 6 | 1781 | |||
AK098751.1 | - | 7 | 1584 | |||
AK130062.1 | - | 6 | 1783 | |||
AK130157.1 | - | 6 | 1782 | |||
AK222925.1 | - | 6 | 1784 | |||
AK223032.1 | - | 6 | 1779 | |||
AK225414.1 | - | 6 | 1616 | |||
AK309997.1 | - | 7 | 1823 | |||
BC001301.1 | - | 6 | 1764 | 3451917 | ||
BC002409.2 | - | 6 | 1769 | 2961617 | ||
BC004251.1 | - | 6 | 1758 | 3623067 | ||
BC008633.1 | - | 5 | 1678 | |||
BC009636.1 | - | 1 | 515 | |||
BC012854.1 | - | 5 | 1654 | |||
BC013380.2 | - | 6 | 1773 | 4025526 | ||
BC013835.1 | - | 6 | 1770 | |||
BC014861.1 | - | 6 | 1771 | 3870487 | ||
BC016045.1 | - | 6 | 1772 | 4564532 | ||
X00351.1 | - | 6 | 1737 | |||
X63432.1 | - | 6 | 1769 |