Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 21 | 2226 | ||
XM_011545742.2 | XP_011544044.1 | 24 | 2436 | |||
XM_011545744.2 | XP_011544046.1 | 9 | 1036 | |||
XM_011545745.1 | XP_011544047.1 | 7 | 804 | |||
XM_011545746.2 | XP_011544048.1 | 7 | 907 | |||
XM_017022947.1 | XP_016878436.1 | 25 | 2509 | |||
XM_017022948.1 | XP_016878437.1 | 8 | 1608 | |||
XM_017022949.1 | XP_016878438.1 | 6 | 1879 | |||
GenBank mRNA | AK098260.1 | - | 19 | 2299 | ||
AK128606.1 | - | 10 | 1094 | |||
AK307544.1 | - | 22 | 2285 | |||
BC109294.1 | - | 9 | 954 | |||
BC109295.1 | - | 9 | 954 |