Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304548.1 | NP_001291477.1 | 61, 62 | 9097 | ||
GenBank mRNA | BG190566.1 | - | 1, 2 | 204 | ||
BG197183.1 | - | 1, 2 | 185 |