Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015094.2 | NP_055909.2 | 3 | 1441 | ||
XM_011530007.2 | XP_011528309.1 | 3 | 3715 | |||
XM_011530008.2 | XP_011528310.1 | 3 | 1631 | |||
XM_011530009.2 | XP_011528311.1 | 4 | 3077 | |||
XM_011530010.2 | XP_011528312.1 | 3 | 1627 | |||
XM_017028669.1 | XP_016884158.1 | 2 | 1344 | |||
GenBank mRNA | AB028943.2 | - | 3 | 1441 | ||
AF349035.2 | - | 1 | 1185 | |||
AL162003.1 | - | 1 | 313 | |||
BC094787.1 | - | 3 | 1374 | 30531041 | ||
CR456377.1 | - | 2 | 1317 |