Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025153.2 | NP_079429.2 | 26 | 4826 | ||
XM_006714833.2 | XP_006714896.1 | 25 | 4407 | |||
XM_011534468.2 | XP_011532770.1 | 25 | 4383 | |||
XM_011534469.1 | XP_011532771.1 | 26 | 4647 | |||
XM_011534472.2 | XP_011532774.1 | 21 | 4123 | |||
XM_017009252.1 | XP_016864741.1 | 25 | 4347 | |||
XM_017009253.1 | XP_016864742.1 | 24 | 4206 | |||
GenBank mRNA | AB018258.2 | - | 26 | 4653 | ||
AK025290.1 | - | 11 | 1749 | |||
AK025294.1 | - | 8 | 1065 |