Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303420.1 | NP_001290349.1 | 10 | 1716 | ||
NM_001303421.1 | NP_001290350.1 | 10 | 1230 | |||
NM_015101.3 | NP_055916.1 | 10 | 1716 | |||
XM_011509332.2 | XP_011507634.1 | 11 | 1522 | |||
XM_011509334.2 | XP_011507636.1 | 11 | 1329 | |||
XM_011509335.2 | XP_011507637.1 | 11 | 1380 | |||
XM_011509336.2 | XP_011507638.1 | 11 | 1157 | |||
XM_011509337.2 | XP_011507639.1 | 7 | 1456 | |||
XM_011509338.2 | XP_011507640.1 | 8 | 1208 | |||
XM_017000750.1 | XP_016856239.1 | 10 | 1281 | |||
XM_017000751.1 | XP_016856240.1 | 5 | 1140 | |||
GenBank mRNA | AB011156.1 | - | 10 | 1641 | ||
AF288389.1 | - | 10 | 1675 | |||
AK095090.1 | - | 5 | 951 | |||
AK095192.1 | - | 10 | 1204 | |||
AK127287.1 | - | 2 | 1221 | |||
AK293973.1 | - | 10 | 1423 | |||
BC035672.1 | - | 10 | 1698 | 5740842 |