Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303420.1 | NP_001290349.1 | 5 | 1195 | ||
NM_001303421.1 | NP_001290350.1 | 5 | 709 | |||
NM_015101.3 | NP_055916.1 | 5 | 1195 | |||
XM_011509332.2 | XP_011507634.1 | 5 | 902 | |||
XM_011509334.2 | XP_011507636.1 | 5 | 709 | |||
XM_011509335.2 | XP_011507637.1 | 5 | 760 | |||
XM_011509336.2 | XP_011507638.1 | 5 | 537 | |||
XM_017000750.1 | XP_016856239.1 | 5 | 760 | |||
GenBank mRNA | AB011156.1 | - | 5 | 1120 | ||
AF288389.1 | - | 5 | 1154 | |||
AK095192.1 | - | 5 | 683 | |||
AK293973.1 | - | 5 | 902 | |||
BC035672.1 | - | 5 | 1177 | 5740842 |