Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000726.3 | NP_000717.2 | 13, 14 | 1370 | ||
NM_001005746.2 | NP_001005746.1 | 13, 14 | 1320 | |||
NM_001005747.2 | NP_001005747.1 | 12, 13 | 1458 | |||
NM_001320722.1 | NP_001307651.1 | 13, 14 | 1618 | |||
XM_011511795.2 | XP_011510097.1 | 13, 14 | 3087 | |||
XM_011511796.2 | XP_011510098.1 | 12, 13 | 2041 | |||
XM_011511797.2 | XP_011510099.1 | 10, 11 | 2090 | |||
XM_017004880.1 | XP_016860369.1 | 10, 11 | 1182 | |||
XM_017004881.1 | XP_016860370.1 | 9, 10 | 1250 | |||
XM_017004882.1 | XP_016860371.1 | 10, 11 | 2575 | |||
XM_017004883.1 | XP_016860372.1 | 14, 15 | 2909 | |||
XM_017004884.1 | XP_016860373.1 | 12, 13 | 2743 | |||
XM_017004885.1 | XP_016860374.1 | 11, 12 | 2583 | |||
XM_017004886.1 | XP_016860375.1 | 4, 5 | 663 | |||
GenBank mRNA | AF038852.1 | - | 13, 14 | 1280 | ||
AK290049.1 | - | 12, 13 | 1439 | |||
AK291378.1 | - | 14, 15 | 1453 | |||
AK294398.1 | - | 13, 14 | 1294 | |||
AK316045.1 | - | 13, 14 | 1311 | |||
AY054985.1 | - | 12, 13 | 1429 | |||
BC075049.2 | - | 13, 14 | 1321 | 30915578 | ||
U95020.1 | - | 13, 14 | 1370 |