Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKLE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015114.2 | NP_055929.1 | 3, 4 | 948 | ||
XM_005266159.3 | XP_005266216.1 | 3, 4 | 869 | |||
XM_005266160.1 | XP_005266217.1 | 3, 4 | 805 | |||
XM_006719735.1 | XP_006719798.1 | 3, 4 | 859 | |||
XM_011534787.2 | XP_011533089.1 | 3, 4 | 859 | |||
XM_011534788.1 | XP_011533090.1 | 3, 4 | 859 | |||
XM_017019076.1 | XP_016874565.1 | 3, 4 | 805 | |||
GenBank mRNA | AB014592.1 | - | 2, 3 | 395 | ||
BC001530.2 | - | 2, 3 | 237 | |||
BC025347.1 | - | 2, 3 | 237 | |||
BC043157.1 | - | 3, 4 | 894 | 5286234 |