Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015123.2 | NP_055938.1 | 22 | 3217 | ||
XM_005264722.1 | XP_005264779.1 | 22 | 2894 | |||
XM_005264723.1 | XP_005264780.1 | 12 | 2376 | |||
XM_017005989.1 | XP_016861478.1 | 22 | 3001 | |||
XM_017005990.1 | XP_016861479.1 | 22 | 2894 | |||
XM_017005991.1 | XP_016861480.1 | 23 | 3054 | |||
XM_017005992.1 | XP_016861481.1 | 22 | 2834 | |||
XM_017005993.1 | XP_016861482.1 | 22 | 2825 | |||
XM_017005994.1 | XP_016861483.1 | 22 | 4667 | |||
XM_017005995.1 | XP_016861484.1 | 16 | 2411 | |||
XM_017005996.1 | XP_016861485.1 | 12 | 2113 | |||
XM_017005997.1 | XP_016861486.1 | 12 | 2376 | |||
GenBank mRNA | AB023230.1 | - | 22 | 2986 | ||
AK024086.1 | - | 6 | 1252 | |||
AK091076.1 | - | 4 | 865 | |||
AK295045.1 | - | 17 | 2459 | |||
AL832231.1 | - | 12 | 2214 | |||
BC028291.1 | - | 2 | 566 | |||
BC110637.1 | - | 7 | 1438 |