Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014839.1 | NP_001014839.1 | 3 | 276 | ||
NM_001014841.1 | NP_001014841.1 | 2 | 208 | |||
NM_014284.2 | NP_055099.1 | 2 | 548 | |||
GenBank mRNA | AB011179.1 | - | 2 | 156 | ||
AB018739.2 | - | 3 | 260 | |||
AB018740.2 | - | 2 | 192 | |||
AL536768.3 | - | 2 | 527 | |||
BC024592.1 | - | 2 | 522 | 3914058 |