Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014839.1 | NP_001014839.1 | 4 | 578 | ||
NM_001014841.1 | NP_001014841.1 | 3 | 510 | |||
NM_014284.2 | NP_055099.1 | 3 | 850 | |||
GenBank mRNA | AB011179.1 | - | 3 | 458 | ||
AB018739.2 | - | 4 | 562 | |||
AB018740.2 | - | 3 | 494 | |||
AL536768.3 | - | 3 | 829 | |||
BC024592.1 | - | 3 | 824 | 3914058 |