Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015135.2 | NP_055950.1 | 6, 7 | 906 | ||
XM_005250235.3 | XP_005250292.1 | 5, 6 | 847 | |||
GenBank mRNA | AK297307.1 | - | 6, 7 | 860 | ||
BC136624.1 | - | 6, 7 | 901 | 9020614 | ||
BC146784.1 | - | 6, 7 | 880 | 9007240 | ||
D86978.1 | - | 6, 7 | 880 |