Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285927.1 | NP_001272856.1 | 27 | 3133 | ||
NM_175291.4 | NP_780500.2 | 27 | 3117 | |||
GenBank mRNA | AK122347.1 | - | 18 | 1971 | ||
BC150753.1 | - | 27 | 3053 | 9087664 | ||
GU797481.1 | - | 27 | 3090 |