Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033606.2 | NP_001028778.2 | 9 | 1488 | ||
NM_001136222.1 | NP_001129694.1 | 8 | 1070 | |||
NM_028817.3 | NP_083093.2 | 10 | 1628 | |||
GenBank mRNA | AK012088.1 | - | 9 | 1431 | ||
AK160839.1 | - | 8 | 1014 | |||
AK161708.1 | - | 10 | 1629 | |||
AK166723.1 | - | 10 | 1566 | |||
BC031529.1 | - | 8 | 1213 | 4009004 |