Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015140.3 | NP_055955.1 | 11 | 1599 | ||
GenBank mRNA | BC001070.2 | - | 11 | 1563 | 3504490 | |
CT841516.1 | - | 11 | 1540 | |||
D63487.1 | - | 11 | 1521 |