Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024413.2 | NP_077724.2 | 2 | 1156 | ||
GenBank mRNA | AK007489.1 | - | 1 | 564 | ||
AK143976.1 | - | 1145 | ||||
AK157990.1 | - | 2 | 1145 | |||
AK170283.1 | - | 2 | 1155 | |||
AK171398.1 | - | 2 | 1145 | |||
BC002120.1 | - | 1 | 1121 | 3491671 |