Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a41b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105159.1 | NP_001098629.1 | 11 | 1216 | ||
GenBank mRNA | BC138083.1 | - | 1145 | 8861102 | ||
BC138084.1 | - | 1143 | 8861103 |
Gene Symbol: Cyp3a41a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017396.3 | NP_059092.2 | 11 | 1212 | ||
GenBank mRNA | AB033414.1 | - | 11 | 1210 |