Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbb-bs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201391.1 | NP_001188320.1 | 2 | 258 | ||
GenBank mRNA | AK002258.1 | - | 2 | 258 | ||
AK003096.1 | - | 2 | 258 | |||
AK003472.1 | - | 2 | 258 | |||
AK005442.1 | - | 2 | 258 | |||
AK005490.1 | - | 2 | 259 | |||
AK010873.1 | - | 2 | 259 | |||
AK010902.1 | - | 2 | 258 | |||
AK010980.1 | - | 2 | 258 | |||
AK010981.1 | - | 2 | 258 | |||
AK010991.1 | - | 2 | 258 | |||
AK010993.1 | - | 2 | 257 | |||
AK011006.1 | - | 2 | 258 | |||
AK011013.1 | - | 2 | 258 | |||
AK011016.1 | - | 2 | 258 | |||
AK011027.1 | - | 2 | 258 | |||
AK011053.1 | - | 2 | 260 | |||
AK011067.1 | - | 2 | 258 | |||
AK011069.1 | - | 2 | 260 | |||
AK011077.1 | - | 2 | 258 | |||
AK011083.1 | - | 2 | 258 | |||
AK011102.1 | - | 2 | 258 | |||
AK027903.1 | - | 2 | 260 | |||
AK027904.1 | - | 2 | 258 | |||
AK088149.1 | - | 2 | 258 | |||
AK133714.1 | - | 2 | 258 | |||
AK147001.1 | - | 2 | 260 | |||
AK160629.1 | - | 2 | 258 | |||
AK161021.1 | - | 2 | 258 | |||
AK165490.1 | - | 2 | 258 | |||
AK168562.1 | - | 2 | 259 | |||
AK168826.1 | - | 2 | 259 | |||
AK168846.1 | - | 2 | 258 |
Gene Symbol: Hbb-b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB020013.1 | - | 2 | 202 | ||
AB364477.1 | - | 2 | 205 |
Gene Symbol: Hbb-bt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008220.5 | NP_032246.2 | 2 | 259 | ||
GenBank mRNA | AK002394.1 | - | 2 | 258 | ||
AK005496.1 | - | 2 | 258 | |||
AK011033.1 | - | 2 | 258 | |||
AK011049.1 | - | 273 | ||||
AK011050.1 | - | 2 | 259 | |||
AK011052.1 | - | 2 | 258 | |||
AK011057.1 | - | 2 | 258 | |||
AK011075.1 | - | 168 | ||||
AK012551.1 | - | 2 | 258 | |||
AK014364.1 | - | 2 | 259 | |||
AK028067.1 | - | 2 | 261 | |||
AK167615.1 | - | 2 | 258 | |||
AK168412.1 | - | 2 | 257 | |||
AK168477.1 | - | 2 | 258 | |||
AK168584.1 | - | 2 | 258 | |||
AK168819.1 | - | 2 | 258 |