Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap26-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027105.2 | NP_081381.1 | 1 | 114 | ||
GenBank mRNA | AK009086.1 | - | 1 | 116 | ||
BC128283.1 | - | 1 | 106 | 40101041 | ||
BC139410.1 | - | 1 | 109 | 8862432 | ||
BC139411.1 | - | 1 | 109 | 8862433 |