Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kazn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109684.1 | NP_001103154.1 | 2 | 703 | ||
NM_001109685.1 | NP_001103155.1 | 3 | 721 | |||
NM_144531.3 | NP_653114.3 | 2 | 703 | |||
GenBank mRNA | AK018497.1 | - | 3 | 699 | ||
AK052809.1 | - | 3 | 722 | |||
AK173090.1 | - | 2 | 703 | |||
BC022941.1 | - | 2 | 707 | |||
CD349067.1 | - | 2 | 278 |