Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM175B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032182.3 | NP_115558.3 | 6 | 533 | ||
XM_011539554.1 | XP_011537856.1 | 5 | 437 | |||
XM_017015986.1 | XP_016871475.1 | 4 | 411 | |||
GenBank mRNA | AK296677.1 | - | 6 | 505 | ||
AL537809.3 | - | 6 | 485 | |||
BC008999.1 | - | 6 | 495 | 2900632 | ||
D63877.1 | - | 6 | 502 | |||
DA148097.1 | - | 6 | 534 |