Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm3417 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123368.1 | NP_001116840.1 | 568 |
Gene Symbol: Gm3448 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123367.1 | NP_001116839.1 | 3 | 568 |
Gene Symbol: Tcte3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011560.3 | NP_035690.2 | 3 | 575 | ||
GenBank mRNA | AK006370.1 | - | 440 | |||
BC048739.1 | - | 504 | ||||
BC061136.1 | - | 458 | 6774834 | |||
BC131981.1 | - | 3 | 566 | 40130258 | ||
BC131983.1 | - | 3 | 566 | 40130260 | ||
BU919334.1 | - | 341 | ||||
U21673.1 | - | 546 | ||||
U21674.1 | - | 3 | 525 |