Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029751.4 | NP_084027.1 | 4 | 449 | ||
GenBank mRNA | AK010983.1 | - | 4 | 420 | ||
AK146543.1 | - | 4 | 419 | |||
AK150820.1 | - | 4 | 418 | |||
AK152611.1 | - | 4 | 419 | |||
AK161942.1 | - | 4 | 419 | |||
AK162047.1 | - | 4 | 420 | |||
AK168258.1 | - | 4 | 419 | |||
BC024609.1 | - | 4 | 393 | 4221597 | ||
BC037146.1 | - | 4 | 461 | 5011713 | ||
CJ042325.1 | - | 4 | 451 |