Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015144.2 | NP_055959.1 | 4 | 401 | ||
XM_005255858.3 | XP_005255915.2 | 4 | 592 | |||
XM_011522964.2 | XP_011521266.1 | 4 | 1495 | |||
XM_017023082.1 | XP_016878571.1 | 4 | 1351 | |||
GenBank mRNA | AB011151.2 | - | 4 | 259 | ||
AB030243.1 | - | 4 | 409 | |||
AK289682.1 | - | 4 | 592 | |||
AL117532.1 | - | 1 | 24 | |||
BC101478.1 | - | 4 | 408 | 8068984 |