Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135577.2 | NP_001129049.1 | 1, 2 | 472 | ||
GenBank mRNA | AK082625.1 | - | 1, 2 | 79 | ||
BC024659.1 | - | 1, 2 | 144 | |||
BC066054.1 | - | 1, 2 | 456 | |||
BC094222.1 | - | 1, 2 | 456 |