Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asphd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039645.1 | NP_001034734.1 | 4 | 424 | ||
GenBank mRNA | AK043553.1 | - | 4 | 619 | ||
AK134834.1 | - | 3 | 1115 | |||
BC111101.1 | - | 4 | 424 | 40049679 |