Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008702.3 | NP_032728.3 | 6 | 2063 | ||
GenBank mRNA | AF036332.1 | - | 6 | 1190 | ||
AK078118.1 | - | 6 | 1172 | |||
BC057667.1 | - | 6 | 1148 | 4235238 | ||
BC058652.1 | - | 6 | 2011 | 6406851 | ||
CB234984.1 | - | 6 | 541 |