Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crygd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007776.2 | NP_031802.2 | 3 | 348 | ||
GenBank mRNA | AK136286.1 | - | 3 | 347 | ||
BC057013.1 | - | 3 | 329 | 6824399 | ||
BC132116.1 | - | 3 | 347 | 40130393 | ||
BC132118.1 | - | 3 | 348 | 40130395 | ||
CA452140.1 | - | 2 | 195 |