Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PASK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252119.1 | NP_001239048.1 | 17 | 3927 | ||
NM_001252120.1 | NP_001239049.1 | 18 | 4059 | |||
NM_001252122.1 | NP_001239051.1 | 17 | 3801 | |||
NM_015148.3 | NP_055963.2 | 17 | 3906 | |||
XM_005246991.1 | XP_005247048.1 | 14 | 3241 | |||
XM_011510826.1 | XP_011509128.1 | 17 | 3882 | |||
XM_011510828.1 | XP_011509130.1 | 18 | 3983 | |||
XM_011510829.1 | XP_011509131.1 | 17 | 3924 | |||
XM_011510830.1 | XP_011509132.1 | 17 | 3925 | |||
XM_011510831.2 | XP_011509133.1 | 15 | 4089 | |||
XM_011510832.2 | XP_011509134.1 | 15 | 4068 | |||
XM_011510833.1 | XP_011509135.1 | 14 | 3262 | |||
XM_011510834.1 | XP_011509136.1 | 10 | 2985 | |||
XM_011510835.1 | XP_011509137.1 | 10 | 2575 | |||
XM_017003634.1 | XP_016859123.1 | 17 | 4150 | |||
XM_017003635.1 | XP_016859124.1 | 18 | 3962 | |||
XM_017003636.1 | XP_016859125.1 | 17 | 3918 | |||
GenBank mRNA | AF387103.1 | - | 17 | 3846 | ||
AK026769.1 | - | 3 | 1554 | |||
AK296945.1 | - | 14 | 3411 | |||
AK302417.1 | - | 17 | 3757 | |||
AK302561.1 | - | 14 | 3241 | |||
BC020054.1 | - | 13 | 3081 | |||
BC043495.1 | - | 11 | 2663 | |||
BC063585.1 | - | 17 | 3849 | 4419998 | ||
CR749231.1 | - | 18 | 4060 | |||
D50925.2 | - | 17 | 3858 | |||
U79240.1 | - | 8 | 1180 |