Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081364.3 | NP_001074833.3 | 18 | 4066 | ||
NM_001128084.2 | NP_001121556.2 | 19 | 4096 | |||
GenBank mRNA | AK122508.1 | - | 1707 | |||
BC076629.1 | - | 18 | 4066 | 30639338 | ||
BC150741.1 | - | 19 | 4025 | 9087652 |