Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023665.3 | NP_076154.3 | 5 | 1044 | ||
GenBank mRNA | AB041555.1 | - | 6 | 1775 | ||
AK003799.1 | - | 4 | 2347 | |||
AK028145.1 | - | 4 | 2349 | |||
AK029529.1 | - | 4 | 2345 | |||
AK150024.1 | - | 5 | 1045 | |||
AK150665.1 | - | 5 | 1024 | |||
AK152736.1 | - | 5 | 1024 | |||
AK160993.1 | - | 4 | 2346 | |||
AK169760.1 | - | 4 | 2347 | |||
AK178549.1 | - | 4 | 390 | |||
AK187384.1 | - | 4 | 392 | |||
BC043057.1 | - | 4 | 2331 | 6419597 | ||
BC046231.1 | - | 4 | 2249 | 6813388 | ||
BC054728.1 | - | 4 | 2319 | 6419723 | ||
BC056986.1 | - | 4 | 2280 | 6816649 |