Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm14430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100415.1 | NP_001093885.1 | 1579 |
Gene Symbol: 0610010B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177543.1 | NP_001171014.1 | 1579 |
Gene Symbol: Gm14305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099327.1 | NP_001092797.1 | 5 | 1571 |
Gene Symbol: Gm14308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099349.2 | NP_001092819.1 | 5 | 1579 |
Gene Symbol: Gm14403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC025828.1 | - | 1 | 522 | ||
BC098094.1 | - | 5 | 774 | 30362243 |
Gene Symbol: Gm14432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100416.2 | NP_001093886.1 | 1571 |
Gene Symbol: Gm14434 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101804.1 | NP_001095274.1 | 1579 |
Gene Symbol: Gm4724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256480.1 | NP_001243409.1 | 1579 |