Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081080.1 | NP_001074549.1 | 3 | 1115 | ||
GenBank mRNA | BC011226.1 | - | 3 | 1110 | ||
BC032205.1 | - | 3 | 1129 | |||
BC137913.1 | - | 3 | 1121 | 8860932 |