Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm8909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081032.2 | NP_001074501.2 | 5 | 943 | ||
GenBank mRNA | AB267095.1 | - | 3 | 561 | ||
AB359231.2 | - | 5 | 943 |