Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pddc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172116.4 | NP_742114.3 | 3, 4 | 307 | ||
GenBank mRNA | AK081187.1 | - | 3, 4 | 267 | ||
AK084273.1 | - | 3, 4 | 263 | |||
AK088159.1 | - | 3, 4 | 283 | |||
AK140975.1 | - | 3, 4 | 258 | |||
AK161205.1 | - | 3, 4 | 273 | |||
BC023835.1 | - | 3, 4 | 277 | 5345186 | ||
BY118625.1 | - | 3, 4 | 307 |