Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024436.3 | NP_077756.2 | 5 | 629 | ||
GenBank mRNA | AB232616.1 | - | 5 | 368 | ||
AJ311124.1 | - | 5 | 425 | |||
AK014407.1 | - | 615 | ||||
AK031667.1 | - | 5 | 609 | |||
AK146594.1 | - | 5 | 629 | |||
AK147619.1 | - | 633 | ||||
AK149200.1 | - | 622 | ||||
AK150617.1 | - | 5 | 635 | |||
BC006596.1 | - | 5 | 616 | 3965264 |