Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004929.3 | NP_004920.1 | 6 | 593 | ||
GenBank mRNA | AK054881.1 | - | 6 | 591 | ||
AK295585.1 | - | 5 | 312 | |||
AK312491.1 | - | 6 | 591 | |||
AK315959.1 | - | 5 | 299 | |||
BC006478.1 | - | 6 | 581 | 2822391 | ||
BC020864.1 | - | 6 | 591 | 4592497 | ||
X06661.1 | - | 6 | 432 |