Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MESDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015154.2 | NP_055969.1 | 2 | 427 | ||
NR_126327.1 | 2 | 427 | ||||
GenBank mRNA | AK026606.1 | - | 2 | 355 | ||
AY359110.1 | - | 2 | 413 | |||
BC009210.2 | - | 2 | 377 | 3637449 | ||
BC012746.2 | - | 2 | 377 | 3954629 | ||
D42039.2 | - | 2 | 343 | |||
DA938708.1 | - | 2 | 427 |