Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsph3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025789.5 | NP_080065.4 | 1 | 308 | ||
GenBank mRNA | AK148230.1 | - | 1 | 270 | ||
BC022760.1 | - | 1 | 268 | |||
BC057649.1 | - | 1 | 267 | |||
BC110424.1 | - | 1 | 243 | |||
CJ173258.1 | - | 1 | 308 |
Gene Symbol: Rsph3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC110284.1 | - | 1 | 229 | 5150198 |