Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271578.1 | NP_001258507.1 | 4 | 1961 | ||
GenBank mRNA | AK030246.1 | - | 4 | 1964 |