Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011563.5 | NP_035693.3 | 2, 3 | 229 | ||
GenBank mRNA | AK008433.1 | - | 2, 3 | 234 | ||
AK011963.1 | - | 177 | ||||
AK014765.1 | - | 1, 2 | 187 | |||
AK088280.1 | - | 2, 3 | 248 | |||
BC002034.1 | - | 2, 3 | 237 | 3485635 | ||
BC081454.1 | - | 2, 3 | 148 | 6818396 | ||
BC086783.1 | - | 1, 2 | 115 | 5700069 | ||
U20611.1 | - | 2, 3 | 191 | |||
U51679.1 | - | 2, 3 | 166 | |||
X82067.1 | - | 2, 3 | 198 |