Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015156.3 | NP_055971.2 | 5 | 816 | ||
GenBank mRNA | AF155595.1 | - | 5 | 579 | ||
BC051003.1 | - | 5 | 814 | |||
BC064495.1 | - | 5 | 814 | |||
D31888.1 | - | 5 | 321 |