Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012518.3 | NP_001012536.2 | 5 | 912 | ||
NM_001109686.2 | NP_001103156.1 | 5 | 891 | |||
NM_001109687.2 | NP_001103157.1 | 5 | 912 | |||
NM_172545.4 | NP_766133.2 | 5 | 891 | |||
GenBank mRNA | AB205007.1 | - | 5 | 918 | ||
AK049454.1 | - | 2 | 162 | |||
AK052174.1 | - | 5 | 874 | |||
AK054555.1 | - | 5 | 859 | |||
AK082062.1 | - | 5 | 902 | |||
AK086833.1 | - | 3 | 777 | |||
BC056938.1 | - | 3 | 613 | |||
BC089302.1 | - | 3 | 759 |