Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019438.1 | NP_062311.1 | 17 | 2679 | ||
GenBank mRNA | AJ237585.1 | - | 11 | 3062 | ||
AK136254.1 | - | 12 | 1665 | |||
AK138026.1 | - | 2 | 1998 | |||
BC137834.1 | - | 10 | 1897 | 8860853 | ||
BC144960.1 | - | 10 | 1897 | 9053508 |